missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ FUK Monoclonal Antibody (6E2)

Description
MA5-15847 targets FUK in indirect ELISA, FACS and WB applications and shows reactivity with Human, mouse, Non-human primate, and Rat samples. The MA5-15847 immunogen is purified recombinant fragment of human FUK expressed in E. Coli. . MA5-15847 detects FUK which has a predicted molecular weight of approximately 117kDa.
The protein encoded by this gene belongs to the GHMP (galacto-, homoserine, mevalonate and phosphomevalonate) kinase family and catalyzes the phosphorylation of L-fucose to form beta-L-fucose 1-phosphate. This enzyme catalyzes the first step in the utilization of free L-fucose in glycoprotein and glycolipid synthesis. L-fucose may be important in mediating a number of cell-cell interactions such as blood group antigen recognition, inflammation, and metastatis. While several transcript variants may exist for this gene, the full-length nature of only one has been described to date.
Specifications
Specifications
| Antigen | FUK |
| Applications | ELISA, Flow Cytometry, Western Blot |
| Classification | Monoclonal |
| Clone | 6E2 |
| Concentration | Conc. Not Determined |
| Conjugate | Unconjugated |
| Formulation | ascites with 0.03% sodium azide |
| Gene | FUK |
| Gene Accession No. | Q7TMC8, Q8N0W3 |
| Gene Alias | 1110046B12Rik; AI303278; Fcsk; Fucokinase; FUK; L-fucose kinase; mFLJ00241 protein; UNQ5838 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?