missing translation for 'onlineSavingsMsg'
Learn More

Melford FUDR

Product Code. 30018271
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30018271 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30018271 Supplier Melford Supplier No. F107051.0
 Restricted Item
Estimated Shipment: 11-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Inhibits DNA synthesis by prior inhibition of thymidylate synthetase.

Inhibits DNA synthesis by prior inhibition of thymidylate synthetase.

Specifications

CAS 50-91-9
Molecular Formula C9H11FN2O5
Quantity 1 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.