missing translation for 'onlineSavingsMsg'
Learn More

Fucosyltransferase 7/FUT7 Antibody (1A12), Novus Biologicals™

Product Code. 18351278 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18351278 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18351278 Supplier Novus Biologicals Supplier No. H00002529M05

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Fucosyltransferase 7/FUT7 Monoclonal antibody specifically detects Fucosyltransferase 7/FUT7 in Human samples. It is validated for ELISA, ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen Fucosyltransferase 7/FUT7
Applications ELISA, Sandwich ELISA
Classification Monoclonal
Clone 1A12
Conjugate Unconjugated
Formulation In 1x PBS, pH 7.4
Gene Accession No. NP_004470
Gene Alias alpha-(1,3)-fucosyltransferase, EC 2.4.1, EC 2.4.1.-, EC 2.4.1.65, Fucosyltransferase 7, fucosyltransferase 7 (alpha (1,3) fucosyltransferase), Fucosyltransferase VII, fuc-TVII, FucT-VII, Galactoside 3-L-fucosyltransferase, Selectin ligand synthase, selectin-ligand synthase
Host Species Mouse
Immunogen FUT7 (NP_004470, 262 a.a. ~ 342 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. YEAFVPADAFVHVDDFGSARELAAFLTGMNESRYQRFFAWRDRLRVRLFTDWRERFCAICDRYPHLPRSQVYEDLEGWFQA
Purification Method IgG purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 2529
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.