missing translation for 'onlineSavingsMsg'
Learn More

Fucosyltransferase 3/FUT3 Mouse anti-Human, Guinea Pig, Clone: 2-25LE; same as LWB01, Novus Biologicals™

Product Code. 18349564 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
20 μg
Unit Size:
100µg
20µg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18349564 20 μg 20µg
18321787 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18349564 Supplier Novus Biologicals Supplier No. NBP31399820UG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Fucosyltransferase 3/FUT3 Monoclonal antibody specifically detects Fucosyltransferase 3/FUT3 in Human, Guinea Pig samples. It is validated for Immunofluorescence, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen Fucosyltransferase 3/FUT3
Applications Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 2-25LE; same as LWB01
Concentration 0.2 mg/ml
Conjugate Unconjugated
Dilution Immunofluorescence 1-2 ug/ml, Immunohistochemistry-Paraffin 1-2 ug/ml
Formulation 10 mM PBS with 0.05% BSA
Gene Accession No. P21217
Gene Alias CD174, FT3B, FucT-III, LE, Les
Host Species Mouse
Immunogen Mucin isolated from a human ovarian cyst fluid (Uniprot: P21217)
Purification Method Protein A or G purified
Quantity 20 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2525
Target Species Human, Guinea Pig
Content And Storage Store at 4°C.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.