missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fucosyltransferase 3/FUT3 Antibody (SPM194) - IHC-Prediluted, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-48372
This item is not returnable.
View return policy
Description
Fucosyltransferase 3/FUT3 Monoclonal specifically detects Fucosyltransferase 3/FUT3 in Human, Guinea Pig samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| Blood Group Lewis b | |
| Monoclonal | |
| Unconjugated | |
| 10mM PBS and 0.05% BSA with 0.05% Sodium Azide | |
| FUT3 | |
| Mucin isolated from a human ovarian cyst fluid (Uniprot: P21217) | |
| Protein A or G purified | |
| RUO | |
| 2525 | |
| Human, Guinea Pig | |
| Purified |
| Immunohistochemistry (Paraffin) | |
| SPM194 | |
| Immunohistochemistry-Paraffin | |
| alpha-(1,3/1,4)-fucosyltransferase, Blood group Lewis alpha-4-fucosyltransferase, CD174, EC 2.4.1, EC 2.4.1.65, FT3B, Fucosyltransferase 3, fucosyltransferase 3 (galactoside 3(4)-L-fucosyltransferase, Lewis blood group), Fucosyltransferase III, FucT-III, galactoside 3(4)-L-fucosyltransferase, LEfucosyltransferase 3 (galactoside 3(4)-L-fucosyltransferase, Lewis blood groupincluded), Les, Lewis FT, MGC131739 | |
| Mouse | |
| 45 kDa | |
| 7 mL | |
| Primary | |
| The Lewis histo-blood group system comprises a set of fucosylated glycosphingolipids that are synthesized by exocrine epithelial cells and circulate in body fluids. The glycosphingolipids function in embryogenesis, tissue differentiation, tumor metastasis, inflammation, and bacterial adhesion. They are secondarily absorbed to red blood cells giving rise to their Lewis phenotype. This gene is a member of the fucosyltransferase family, which catalyzes the addition of fucose to precursor polysaccharides in the last step of Lewis antigen biosynthesis. It encodes an enzyme with alpha(1,3)-fucosyltransferase and alpha(1,4)-fucosyltransferase activities. Lewis blood group antigens are carbohydrate moieties structurally integrated in mucous secretions. Lewis antigen system alterations have been described in gastric carcinoma and associated lesions. Anomalous expression of Lewis B antigen has been found in some non-secretory gastric carcinomas and colorectal cancers. | |
| Store at 4C. | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction