missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fucosyltransferase 3/FUT3 Antibody (2-25LE; same as LWB01) [Janelia Fluor™ 669], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP3-14260JF669
This item is not returnable.
View return policy
Description
Fucosyltransferase 3/FUT3 Monoclonal antibody specifically detects Fucosyltransferase 3/FUT3 in Guinea Pig,Human samples. It is validated for Immunohistochemistry (Paraffin),Immunocytochemistry/ Immunofluorescence
Specifications
| Fucosyltransferase 3/FUT3 | |
| Monoclonal | |
| Janelia Fluor 669 | |
| CD174, FT3B, FucT-III, LE, Les | |
| Mucin isolated from a human ovarian cyst fluid (Uniprot: P21217) | |
| 0.1 mL | |
| Primary | |
| Guinea Pig, Human | |
| Purified |
| Immunohistochemistry (Paraffin), Immunocytochemistry/Immunofluorescence | |
| 2-25LE; same as LWB01 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| 2525 | |
| Store at 4°C in the dark. | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction