missing translation for 'onlineSavingsMsg'
Learn More

Fucosyltransferase 3/FUT3 Antibody (2-25LE; same as LWB01), DyLight 350, Novus Biologicals™

Product Code. 18344087 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18344087 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18344087 Supplier Novus Biologicals Supplier No. NBP314260UV

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Fucosyltransferase 3/FUT3 Monoclonal antibody specifically detects Fucosyltransferase 3/FUT3 in Human, Guinea Pig samples. It is validated for Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen Fucosyltransferase 3/FUT3
Applications Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 2-25LE; same as LWB01
Conjugate DyLight 350
Dilution Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate
Gene Alias CD174, FT3B, FucT-III, LE, Les
Host Species Mouse
Immunogen Mucin isolated from a human ovarian cyst fluid (Uniprot: P21217)
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2525
Target Species Human, Guinea Pig
Content And Storage Store at 4°C in the dark.
Product Type Antibody
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.