missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fucosyltransferase 3/FUT3 Antibody (2-25LE), mFluor Violet 500 SE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB500-526MFV500
This item is not returnable.
View return policy
Description
Fucosyltransferase 3/FUT3 Monoclonal antibody specifically detects Fucosyltransferase 3/FUT3 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| Fucosyltransferase 3/FUT3 | |
| Monoclonal | |
| mFluor Violet 500 SE | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| 2525 | |
| Store at 4C in the dark. | |
| IgG1 |
| Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| 2-25LE | |
| Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin | |
| CD174, FT3B, FucT-III, LE, Les | |
| Mucin isolated by size exclusion chromatography from an ovarian cyst fluid (pure endocervical type according to the Fenoglio's classification). | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction