missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fucosyltransferase 2/FUT2 Rabbit anti-Human, Mouse, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-03645-100UL
This item is not returnable.
View return policy
Description
Fucosyltransferase 2/FUT2 Polyclonal specifically detects Fucosyltransferase 2/FUT2 in Human, Mouse samples. It is validated for Western Blot.
Specifications
| Fucosyltransferase 2/FUT2 | |
| Polyclonal | |
| PBS with 50% glycerol, pH7.3. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 2524 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| alpha (1,2) fucosyltransferase, Alpha(1,2)FT 2, alpha(1,2)FT2, B12QTL1, EC 2.4.1.69, Fucosyltransferase 2, fucosyltransferase 2 (secretor status included), galactoside 2-alpha-L-fucosyltransferase 2, GDP-L-fucose:beta-D-galactoside 2-alpha-L-fucosyltransferase 2, Se, SE2, SEC2SE, Secretor blood group alpha-2-fucosyltransferase, Secretor factor, sej | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 29-150 of human Fucosyltransferase 2/FUT2 (NP_001091107.1). | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction