missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fucosyltransferase 1/FUT1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-35576-20ul
This item is not returnable.
View return policy
Description
Fucosyltransferase 1/FUT1 Polyclonal antibody specifically detects Fucosyltransferase 1/FUT1 in Human,Mouse,Rat samples. It is validated for ELISA,Western Blot
Specifications
| Fucosyltransferase 1/FUT1 | |
| Polyclonal | |
| Western Blot 1:500 - 1:2000, ELISA | |
| alpha (1,2) fucosyltransferase, Alpha(1,2)FT 1,2-alpha-L-fucosyltransferase, Blood group H alpha 2-fucosyltransferase, EC 2.4.1.69, Fucosyltransferase 1, fucosyltransferase 1 (galactoside 2-alpha-L-fucosyltransferase), fucosyltransferase 1 (galactoside 2-alpha-L-fucosyltransferase, Bombayphenotype included), fucosyltransferase 1 (galactoside 2-alpha-L-fucosyltransferase, H blood group), galactoside 2-alpha-L-fucosyltransferase 1, GDP-L-fucose:beta-D-galactoside 2-alpha-L-fucosyltransferase 1, HHH, HSC | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 130-310 of human Fucosyltransferase 1/FUT1 (NP_000139.1).,, Sequence:, VLAPEVDSRTPWRELQLHDWMSEEYADLRDPFLKLSGFPCSWTFFHHLREQIRREFTLHDHLREEAQSVLGQLRLGRTGDRPRTFVGVHVRRGDYLQVMPQRWKGVVGDSAYLRQAMDWFRARHEAPVFVVTSNGMEWCKENIDTSQGDVTFAGDGQEATPWKDFALLTQCNHTIMTIGTF | |
| 20 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| ELISA, Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 2523 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction