missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ FUCA2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5112849
This item is not returnable.
View return policy
Description
FUCA2 Polyclonal Antibody for Western Blot, IHC (P), ELISA
FUCA2 (Alpha-L-fucosidase) catalyzes the hydrolysis of terminal alpha-L-fucosidase linkages in glycosphingolipids and glycoproteins. At least 2 separate polymorphic alpha-L-fucosidases are recognized in man. The FUCA2 locus regulates the level of alpha-fucosidase in plasma and fibroblasts but not in leukocytes. In fucosidosis, deficiency of alpha-L-fucosidase is found in both plasma and leukocytes.
Specifications
| FUCA2 | |
| Polyclonal | |
| Unconjugated | |
| FUCA2 | |
| 0610025O11Rik; 5530401P20Rik; Alpha-L-fucosidase 2; alpha-L-fucoside fucohydrolase 2; dJ20N2.5; fc20c06; Fuca2; fucosidase, alpha-L- 2, plasma; MGC1314; plasma alpha-L-fucosidase; PSEC0151; UNQ227/PRO260; wu:fc20c06; zgc:92013 | |
| Rabbit | |
| Protein G | |
| RUO | |
| 2519 | |
| -20°C or -80°C if preferred | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Western Blot | |
| 0.75 mg/mL | |
| PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
| Q9BTY2 | |
| FUCA2 | |
| Recombinant Human Plasma alpha-L-fucosidase protein (29-465AA). | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction