missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ FUBP3 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA583874
This item is not returnable.
View return policy
Description
Immunogen sequence: GTNLGAPGAF GQSPFSQPPA PPHQNTFPPR SSGCFPNMAA KVNGNPHSTP VSGPPAFLTQ GWGSTYQAWQ QPTQQVPSQQ SQPQSSQPNY SK Highest antigen sequence indentity to the following orthologs: Mouse - 88%, Rat - 88%.
May interact with single-stranded DNA from the far-upstream element (FUSE). May activate gene expression.
Specifications
| FUBP3 | |
| Polyclonal | |
| Unconjugated | |
| FUBP3 | |
| A330051M14Rik; AV006371; far upstream element (FUSE) binding protein 3; far upstream element binding protein 3; far upstream element-binding protein 3; FBP3; FUBP3; FUSE-binding protein 3; MAP2 RNA trans-acting protein 2; Marta2 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 8939 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q96I24 | |
| FUBP3 | |
| Recombinant protein corresponding to Human FUBP3. Recombinant protein control fragment (Product #RP-100913). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction