missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FUBP3 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-38488-20ul
This item is not returnable.
View return policy
Description
FUBP3 Polyclonal antibody specifically detects FUBP3 in Human,Mouse samples. It is validated for ELISA,Western Blot
Specifications
| FUBP3 | |
| Polyclonal | |
| Western Blot 1:500 - 1:2000, ELISA | |
| far upstream element (FUSE) binding protein 3, far upstream element-binding protein 3, FBP3, FLJ25229, FUSE-binding protein 3 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 65-260 of human FUBP3 (NP_003925.1).,, Sequence:, GNQLGALVHQRTVITEEFKVPDKMVGFIIGRGGEQISRIQAESGCKIQIASESSGIPERPCVLTGTPESIEQAKRLLGQIVDRCRNGPGFHNDIDSNSTIQEILIPASKVGLVIGRGGETIKQLQERTGVKMVMIQDGPLPTGADKPLRITGDAFKVQQAREMVLEIIREKDQADFRGVRGDFNSRMGGGSIEVSV | |
| 20 μL | |
| Primary | |
| Human, Mouse | |
| Purified |
| ELISA, Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 8939 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction