missing translation for 'onlineSavingsMsg'
Learn More

FUBP3 Antibody, DyLight 755, Novus Biologicals™

Product Code. 18406642 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18406642 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18406642 Supplier Novus Biologicals Supplier No. NBP150045IR

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

FUBP3 Polyclonal antibody specifically detects FUBP3 in Human, Mouse samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen FUBP3
Applications Western Blot
Classification Polyclonal
Conjugate DyLight 755
Dilution Western Blot
Formulation 50mM Sodium Borate
Gene Alias far upstream element (FUSE) binding protein 3, far upstream element-binding protein 3, FBP3, FLJ25229, FUSE-binding protein 3
Host Species Rabbit
Immunogen The immunogen for this product maps to a region between residue 522 and 572 of human FUBP3 using the numbering given in entry NP_003925.1 (GeneID 8939).
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8939
Target Species Human, Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.