missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ FUBP1 Polyclonal Antibody

Goat Polyclonal Antibody
Brand: Invitrogen™ PA5142922
This item is not returnable.
View return policy
Description
This antibody is tested in Peptide ELISA: antibody detection limit dilution 1:128,000.
This gene encodes a ssDNA binding protein that activates the far upstream element (FUSE) of c-myc and stimulates expression of c-myc in undifferentiated cells. Regulation of FUSE by FUBP occurs through single-strand binding of FUBP to the non-coding strand. This protein has been shown to function as an ATP-dependent DNA helicase.
Specifications
| FUBP1 | |
| Polyclonal | |
| Unconjugated | |
| FUBP1 | |
| 9530027K12Rik; D3Ertd330e; DNA helicase V; EMBL:AAI07943.1, ECO:0000312; EMBL:AAI07943.1}; far upstream element (FUSE) binding protein 1; far upstream element (FUSE) binding protein 4; far upstream element binding protein 1; far upstream element-binding protein; far upstream element-binding protein 1; far upstream element-binding protein 1 {ECO:0000250; FBP; FBP {ECO:0000250; Fubp; Fubp1; fubp1 {ECO:0000312; Fubp4; FUSE binding protein 1; FUSE-binding protein 1; FUSE-binding protein 1 {ECO:0000250; hDH V; RGD:1591892}; UniProtKB:Q96AE4, ECO:0000312; UniProtKB:Q96AE4} | |
| Goat | |
| Ammonium sulfate precipitation | |
| RUO | |
| 8880 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.5 mg/mL | |
| TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
| Q96AE4 | |
| FUBP1 | |
| Peptide with sequence C-DQIVEKGRPAPGFHH. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction