missing translation for 'onlineSavingsMsg'
Learn More

FUBI/MNSF beta/FAU Antibody (3C10), Novus Biologicals™

Product Code. 18394548 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18394548 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18394548 Supplier Novus Biologicals Supplier No. H00002197M03

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

FUBI/MNSF beta/FAU Monoclonal antibody specifically detects FUBI/MNSF beta/FAU in Human, Squirrel samples. It is validated for Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen FUBI/MNSF beta/FAU
Applications Western Blot, ELISA, Immunocytochemistry
Classification Monoclonal
Clone 3C10
Conjugate Unconjugated
Dilution Western Blot 1:500, ELISA, Immunocytochemistry/ Immunofluorescence
Formulation In 1x PBS, pH 7.4
Gene Accession No. AAH33877
Gene Alias 40S ribosomal protein S30, asr1, FAU1, FAU-encoded ubiquitin-like protein, FBR-MuSV-associated ubiquitously expressed, Finkel-Biskis-Reilly murine sarcoma virus (FBR-MuSV) ubiquitously expressed, Finkel-Biskis-Reilly murine sarcoma virus (FBR-MuSV) ubiquitously expressed(fox derived), FLJ22986, Fub1, Fubi, MNSFbeta, monoclonal nonspecific suppressor factor beta, ribosomal protein S30, RPS30, ubiquitin-like protein fubi and ribosomal protein S30, ubiquitin-like-S30 fusion protein
Host Species Mouse
Immunogen FAU (AAH33877, 1 a.a. ~ 133 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. MQLFVRAQELHTFEVTGQETVAQIKAHVASLEGIAPEDQVVLLAGAPLEDEATLGQCGVEALTTQEVAGRMLGGKVHGSLARAGKVRGQTPKVAKQEKKKKKTGRAKRRMQYNRRFVNVVPTFGKKKGPNANS
Purification Method IgG purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2197
Target Species Human, Squirrel
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.