missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ FTSJD2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA544281
This item is not returnable.
View return policy
Description
Peptide sequence: YRLEEMEKIF VRLEMKIIKG SSGTPKLSYT GRDDRHFVPM GLYIVRTVNE Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 93%; Horse: 100%; Human: 100%; Mouse: 100%; Pig: 100%; Rabbit: 93%; Rat: 100%; Zebrafish: 86%.
S-adenosyl-L-methionine-dependent methyltransferase that mediates mRNA cap1 2'-O-ribose methylation to the 5'-cap structure of mRNAs. Methylates the ribose of the first nucleotide of a m(7)GpppG-capped mRNA and small nuclear RNA (snRNA) to produce m(7)GpppRm (cap1). Displays a preference for cap0 transcripts. Cap1 modification is linked to higher levels of translation. May be involved in the interferon response pathway. [UniProt].
Specifications
| FTSJD2 | |
| Polyclonal | |
| Unconjugated | |
| CMTR1 | |
| 1300018I05Rik; AI451264; Cap methyltransferase 1; Cap1 2'O-ribose methyltransferase 1; cap-specific mRNA (nucleoside-2'-O-)-methyltransferase 1; cmtr1; FtsJ methyltransferase domain containing 2; ftsJ methyltransferase domain-containing protein 2; ftsjd2; hMTr1; hypothetical protein LOC393102; interferon-stimulated gene 95 kDa protein; ISG95; KIAA0082; LOW QUALITY PROTEIN: cap-specific mRNA (nucleoside-2'-O-)-methyltransferase 1; LRRG00129; mKIAA0082; MTr1; RGD1307801; S-adenosyl-L-methionine-dependent methyltransferase ftsjd2; zgc:55336 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 23070 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q8N1G2 | |
| CMTR1 | |
| Synthetic peptide directed towards the C-terminal of human FTSJD2 (aa 707-756). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction