missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ FTSJD1 Polyclonal Antibody
GREENER_CHOICE

Product Code. 15777531 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15777531 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15777531 Supplier Invitrogen™ Supplier No. PA559681

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Immunogen sequence: AIHPLLSKMT LNFGTEMKRK ALFPHHVIPD SFLKRHEECC VFFHKYQLET ISENIRLFEC MGKAEQEKLN NLRDCAIQYF MQKFQLKHLS RNNWLVK Highest antigen sequence identity to the following orthologs: Mouse - 84%, Rat - 81%.

S-adenosyl-L-methionine-dependent methyltransferase that mediates mRNA cap2 2'-O-ribose methylation to the 5'-cap structure of mRNAs. Methylates the ribose of the second nucleotide of a m(7)GpppG-capped mRNA and small nuclear RNA (snRNA) (cap0) to produce m(7)GpppRmpNm (cap2). Recognizes a guanosine cap on RNA independently of its N(7) methylation status. Display cap2 methylation on both cap0 and cap1. Displays a preference for cap1 RNAs.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FTSJD1
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene CMTR2
Gene Accession No. Q8IYT2
Gene Alias adrift homolog; AFT; AU022703; C730036L12Rik; cap methyltransferase 2; cap2 2'O-ribose methyltransferase 2; Cap-specific mRNA (nucleoside-2'-O-)-methyltransferase 2; Cmtr2; FtsJ methyltransferase domain containing 1; ftsJ methyltransferase domain-containing protein 1; ftsjd1; HMTr2; hypothetical protein LOC406742; MTr2; Protein adrift homolog; RGD1309394; wu:fj48c05; zgc:55686
Gene Symbols CMTR2
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human FTSJD1. Recombinant protein control fragment (Product #RP-98779).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55783
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.