missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FTSJ1 Antibody, DyLight 680, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-26088FR
This item is not returnable.
View return policy
Description
FTSJ1 Polyclonal antibody specifically detects FTSJ1 in Human samples. It is validated for Western Blot, Immunocytochemistry
Specifications
| FTSJ1 | |
| Polyclonal | |
| Western Blot, Immunocytochemistry | |
| CDLIV, cell division protein, EC 2.1.1.-, FtsJ homolog 1 (E. coli), JM23, mental retardation, X-linked 44, mental retardation, X-linked 9, MRX44, Protein ftsJ homolog 1, putative ribosomal RNA methyltransferase 1, rRNA (uridine-2'-O-)-methyltransferase, SPB1MRX9, TRM7 | |
| Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Human FTSJ1 | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Immunocytochemistry | |
| DyLight 680 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| 24140 | |
| Store at 4C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction