missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FTS Antibody, DyLight 650, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-97179C
This item is not returnable.
View return policy
Description
FTS Polyclonal antibody specifically detects FTS in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| FTS | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Immunohistochemistry (Paraffin) | |
| DyLight 650 | |
| AKT interacting protein, AKT-interacting protein, FLJ13258, Ft1, FTSfused toes homolog (mouse), fused toes (mouse) homolog, fused toes homolog, Fused toes protein homolog | |
| Produced in rabbits immunized with E. coli-derived Human FTS fragment. | |
| 0.1 mL | |
| Apoptosis | |
| 64400 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction