missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FTO, Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit Polyclonal Antibody
Brand: Abnova PAB12425.100uL
This item is not returnable.
View return policy
Specifications
| FTO | |
| Polyclonal | |
| Western Blot (0.5 ug/mL) The optimal working dilution should be determined by the end user. | |
| fat mass and obesity associated | |
| FTO | |
| A synthetic peptide corresponding to amino acids 400-505 of human FTO. | |
| RUO | |
| 79068 | |
| Store at 4°C for short term. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. | |
| Liquid |
| Western Blot | |
| Unconjugated | |
| In Tris-citrate/phosphate buffer, pH 7.0-8.0 (0.09% sodium azide) | |
| KIAA1752/MGC5149 | |
| Rabbit | |
| 100 μL | |
| Primary | |
| Human, Mouse, Primate | |
| Antibody |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction