missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FTO, Mouse anti-Human,Mouse,Rat, Clone: FT86-4, Novus Biologicals™
Mouse Monoclonal Antibody
£277.00 - £395.00
Specifications
| Antigen | FTO |
|---|---|
| Clone | FT86-4 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot, ELISA, Immunoprecipitation |
| Applications | Western Blot, ELISA, Immunoprecipitation |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18271138
|
Novus Biologicals
NBP2-80045-50UG |
50 μg |
£277.00
50µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18244428
|
Novus Biologicals
NBP2-80045-100UG |
100 μg |
£395.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
FTO Monoclonal specifically detects FTO in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunoprecipitation.Specifications
| FTO | |
| 1 mg/mL | |
| Western Blot, ELISA, Immunoprecipitation | |
| Unconjugated | |
| Mouse | |
| Lipid and Metabolism, Neuroscience | |
| 0.2 um-filtered solution in PBS, pH 7.4 | |
| 79068 | |
| Recombinant human FTO. | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| FT86-4 | |
| Western Blot, ELISA, Immunoprecipitation | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| alpha-ketoglutarate-dependent dioxygenase FTO, EC 1.14.11.-, fat mass and obesity associated, Fat mass and obesity-associated protein, KIAA1752protein fto, MGC5149 | |
| FTO | |
| IgG1 κ | |
| Protein G purified | |
| Recognizes human, mouse and rat FTO. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title