missing translation for 'onlineSavingsMsg'
Learn More

FTO Antibody [mFluor Violet 500 SE], Novus Biologicals™

Product Code. 30732483 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30732483 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30732483 Supplier Novus Biologicals Supplier No. AF7208MFV500

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Sheep Polyclonal Antibody

FTO Polyclonal antibody specifically detects FTO in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen FTO
Applications Western Blot
Classification Polyclonal
Conjugate mFluor Violet 500 SE
Formulation 50mM Sodium Borate
Gene Alias alpha-ketoglutarate-dependent dioxygenase FTO, EC 1.14.11.-, fat mass and obesity associated, Fat mass and obesity-associated protein, KIAA1752protein fto, MGC5149
Host Species Sheep
Immunogen E. coli-derived recombinant human FTO, Met1-Arg178, Accession # Q9C0B1
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Lipid and Metabolism, Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 79068
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.