Learn More
Specac™ FTIR Polariser Mount With Slide Mounted Plate and Rotating Ring
Product Code. 15210158
Encompass_Preferred
Click to view available options
Type:
FTIR Polariser Mount
Product Code. 15210158
Brand: Specac™ GS12500
Polarizer Rotator with 3” x 2” Slide Mount for FTIR Infrared Polarizers. Can be used in all FTIR instruments.
Specifications
Type FTIR Polariser Mount
Description 3 x 2 in. Slide Mount for FTIR Infrared Polarizers