missing translation for 'onlineSavingsMsg'
Learn More

Ftaxilide, MedChemExpress

Product Code. 16408658
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16408658 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16408658 Supplier Medchem Express Supplier No. HYB1040
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

MedChemExpress Ftaxilide is a novel antituberculosis agent.

Ftaxilide is a novel antituberculosis agent.

Specifications

Chemical Name or Material Ftaxilide
Physical Form Solid
CAS 19368-18-4
Color White
Quantity 10 mg
Purity Grade Notes Research
Molecular Formula C16H15NO3
For Use With (Application) COVID-19-immunoregulation
Solubility Information DMSO : ≥ 40 mg/mL (148.53 mM)
SMILES O=C(O)C1=CC=CC=C1C(NC2=C(C)C=CC=C2C)=O
Molecular Weight (g/mol) 269.3
Formula Weight 269.3
Percent Purity 99.17%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.