missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Specac™ FT-IR Rotatable Polarizer Mount 50mm
Click to view available options
Type:
FT-IR Rotatable Polarizer Mount 50mm
Description
This rotatable polarizer mount for 50.0mm diameter clear aperture Polarizers are graduated in 2 degree angle intervals and can be estimated to 1 degree accuracy. They can be mounted directly either to a bench or via a support post.
Specifications
Specifications
| Type | FT-IR Rotatable Polarizer Mount 50mm |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction