Learn More
Specac™ FT-IR Research Starter Kit plus Gas Pack Add-On
Product Code. 15219827
Encompass_Preferred
Change view
Click to view available options
Type:
FT-IR Research Starter Kit
Product Code. Type
15219827 FT-IR Research Starter Kit
1 options
Product Code. 15219827 Supplier Specac™ Supplier No. GS01191
Estimated Shipment: 25-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
This kit includes a Quest ATR ZnSe accessory with liquid cell , KBR + CaF2 windows, spacers, nujol, fluorolube, luer syringe, 2 ton mini-pellet press, 7mm die, pellet ring holder, pestle and mortar, KBr powder and mount as well as Storm 10cm pyrex gas cell with KBr windows, mount and seal kit.
Specifications
Type FT-IR Research Starter Kit
Includes A Quest ATR ZnSe accessory with liquid cell , KBR + CaF2 windows, spacers, nujol, fluorolube, luer syringe, 2 ton mini-pellet press, 7 mm die, pellet ring holder, pestle and mortar, KBr powder and mount as well as Storm 10 cm pyrex gas cell with KBr windows, mount and seal kit.