Learn More
Specac™ FT-IR Oil in Water Analysis Kit
Description
This kit is designed for the analysis of total recoverable oil and grease or petroleum hydrocarbons in water. This kit contains matched pairs of infrared grade quartz cells supplied in 3 different pathlengths of 10mm, 50mm and 100mm. They are all stoppered to ensure retention of sample and include a 3in. x 2in. mount for use in any FTIR instrument.
Specifications
Specifications
| Type | FT-IR Oil in Water Analysis Kit |
| Includes | Matched pairs of infrared grade quartz cells supplied in 3 different pathlengths of 10 mm, 50 mm and 100 mm |