Learn More
Specac™ FT-IR Liquid Pack Complete
Product Code. 15239817
Encompass_Preferred
Change view
Click to view available options
Type:
FT-IR Liquid Pack Complete
Product Code. Type
15239817 FT-IR Liquid Pack Complete
1 options
Product Code. 15239817 Supplier Specac™ Supplier No. GS01140
Estimated Shipment: 19-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
The versatile Omni-Cell system is at the core of the liquid pack which contains all the essential accessories needed for the analysis of liquids, gums and pastes including a Syringe, KBr windows, CaF2 windows, PTFE spacers, Nujol and Fluorolube.
Specifications
Type FT-IR Liquid Pack Complete