Learn More
Specac™ FT-IR Basic Starter Kit plus Gas Pack Add-On
Click to view available options
Type:
FT-IR Basic Starter Kit
Description
This kit includes a mini-pellet press, 7mm pellet die, KBr powder, pestle and mortar, ring holder and mount as well as the Omni-Cell system, KBr windows, CaF2 windows, PTFE Spacers, nujol, fluorolube and Pyrex 10cm Gas Cell
Specifications
Specifications
| Type | FT-IR Basic Starter Kit |
| Includes | A mini-pellet press, 7 mm pellet die, KBr powder, pestle and mortar, ring holder and mount as well as the Omni-Cell system, KBr windows, CaF2 windows, PTFE Spacers, nujol, fluorolube and Pyrex 10 cm Gas Cell |