Learn More
Specac™ FT-IR Basic Starter Kit
Product Code. 15269817
Encompass_Preferred
Change view
Click to view available options
Type:
FT-IR Basic Starter Kit
Product Code. Type
15269817 FT-IR Basic Starter Kit
1 options
Product Code. 15269817 Supplier Specac™ Supplier No. GS01180
Estimated Shipment: 04-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
This kit includes a mini-pellet press, 7mm pellet die, KBr powder, pestle and mortar, ring holder and mount as well as the Omni-Cell system, KBr windows, CaF2 windows, PTFE Spacers, nujol and fluorolube.
Specifications
Type FT-IR Basic Starter Kit
Includes A mini-pellet press, 7 mm pellet die, KBr powder, pestle and mortar, ring holder and mount as well as the Omni-Cell system, KBr windows, CaF2 windows, PTFE Spacers, nujol and fluorolube