Learn More
Specac™ FT-IR Analyst Starter Kit plus Gas Pack Add-On
Description
This kit includes a liquid cell with KBr windows, CaF2 windows, spacers as well as a 15T manual hydraulic press, 13mm die, pestle and mortar, KBr powder, disc holder and mount, plus storm 10cm pyrex gas cell with KBr windows, mount and seal kit.
Specifications
Specifications
| Type | FT-IR Analyst Starter Kit |
| Includes | A liquid cell with KBr windows, CaF2 windows, spacers as well as a 15 T manual hydraulic press, 13 mm die, pestle and mortar, KBr powder, disc holder and mount, plus storm 10 cm pyrex gas cell with KBr windows, mount and seal kit. |