Learn More
Specac™ FT-IR Advanced Starter Kit plus Gas Pack Add-On
Product Code. 15239827
Encompass_Preferred
Click to view available options
Type:
FT-IR Advanced Starter Kit
Product Code. 15239827
Brand: Specac™ GS01196
This kit includes a Quest ATR ZnSe accessory with liquid cell, KBR + CaF2 windows, spacers, nujol, fluorolube, luer syringe, 15 ton manual hydraulic press, pestle 7 mortar, KBr powder, 13mm dis holder with mount and 13mm evacuable pellet die as well as a Storm 10cm pyrex gas cell with KBr windows, mount and seal kit.
Specifications
Type FT-IR Advanced Starter Kit