Learn More
Specac™ FT-IR Advanced Solid Pack Complete
Product Code. 15249817
Encompass_Preferred
Change view
Click to view available options
Type:
FT-IR Advanced Solid Pack Complete
Product Code. Type
15249817 FT-IR Advanced Solid Pack Complete
1 options
Product Code. 15249817 Supplier Specac™ Supplier No. GS01160
Estimated Shipment: 21-10-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
The 15T Manual Hydraulic Press is central to this Pack which also includes a Pestle and Mortar, KBr Powder, 13mm Disc Holder with Mount and 13mm Evacuable Pellet Die
Specifications
Type FT-IR Advanced Solid Pack Complete
Includes A Pestle and Mortar, KBr Powder, 13 mm Disc Holder with Mount and 13 mm Evacuable Pellet Die