Learn More
FSTL3 Rabbit, anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 28218-1-AP-20UL
Description
The FSTL3 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human FSTL3. This antibody recognizes human, mouse, rat antigen. The FSTL3 antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Specifications
| FSTL3 | |
| 600 μg/mL | |
| Rabbit | |
| Primary | |
| Human, Mouse, Rat | |
| 150 kDa | |
| IgG |
| Western Blot, Immunohistochemistry, ELISA | |
| FLRG, Follistatin like protein 3, Follistatin related protein 3, FSRP, FSTL3 | |
| 20 μL | |
| 10272 | |
| Store at -20°C. Stable for one year after shipment. | |
| Liquid |