missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ FSIP1 Monoclonal Antibody (OTI4F9)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA526776
This item is not returnable.
View return policy
Description
FSIP1 Monoclonal Antibody for Western Blot, IHC (P)
The exact functions of FSIP1 remain unknown.
Specifications
| FSIP1 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q8NA03 | |
| FSIP1 | |
| Full length human recombinant protein of FSIP1 produced in HEK293T cell. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI4F9 | |
| Unconjugated | |
| FSIP1 | |
| 1700012M13Rik; 4933432K11Rik; fibrous sheath interacting protein 1; fibrous sheath-interacting protein 1; Fsip1; HSD10 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 161835 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction