Learn More
Invitrogen™ FSHR Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA595380
Description
Human FSH Receptor shares 91.2% and 90% amino acid (aa) sequence identity with mouse and rat FSH Receptor, respectively. Reconstitute with 0.2 mL of distilled water to yield a concentration of 500 μg/mL. Positive Control - WB: rat testis tissue, rat ovary tissue, mouse testis tissue, mouse ovary tissue, HELA whole cell. Store at -20°C for one year from date of receipt. After reconstitution, at 4°C for one month. It can also be aliquotted and stored frozen at -20°C for six months. Avoid repeated freeze-thaw cycles.
Follicle-Stimulating Hormone Receptor (FSHR) expression has been reported in ovary, uterus, and testis. ESTs have been isolated from testis libraries. G-protein Coupled Receptors (GPCRs) comprise one of the largest families of signaling molecules with more than a thousand members currently predicted to exist. All GPCRs share a structural motif consisting of seven membrane-spanning helices, and exist in both active and inactive forms. An array of activating ligands participate in the conformation of GPCRs which leads to signaling via G-proteins and downstream effectors. Ongoing studies have also shown the vast series of reactions which participate in the negative regulation of GPCRs. This turn-off activity has tremendous implications for the physiological action of the cell, and continues to drive pharmacological research for new drug candidates. Two blockbuster drugs which have been developed as GPCR-targeted pharmaceuticals are Zyprexa (Eli Lilly) and Claritin (Schering-Plough) which have multi-billion dollar shares of the mental health and allergy markets, respectively.
Specifications
| FSHR | |
| Polyclonal | |
| Unconjugated | |
| FSHR | |
| follicle stimulating hormone receptor; follicle-stimulating hormone receptor; Follitropin receptor; FSH receptor; Fshr; FSH-R; FSHR1; FSHRO; LGR1; ODG1 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 14309, 2492, 25449 | |
| -20°C | |
| Lyophilized |
| Western Blot | |
| 500 μg/mL | |
| PBS with 5mg BSA and 0.05mg sodium azide | |
| P20395, P23945, P35378 | |
| FSHR | |
| E.coli-derived human FSH Receptor recombinant protein (Position: C18-N187). | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.