missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FSHR Antibody (6E8.2F5), DyLight 650, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-36489C
This item is not returnable.
View return policy
Description
FSHR Monoclonal specifically detects FSHR in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| FSHR | |
| Monoclonal | |
| DyLight 650 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| follicle stimulating hormone receptor, follicle-stimulating hormone receptor, Follitropin receptor, FSH receptor, FSH-R, LGR1FSHRO, MGC141667, MGC141668, ODG1 | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| 6E8.2F5 | |
| Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin | |
| P23945 | |
| FSHR | |
| A synthetic peptide made to the human FSH R protein sequence (between residues 1-200). [Swiss-Prot P23945] | |
| 0.1 mL | |
| GPCR | |
| 2492 | |
| Store at 4C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction