missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FSH-beta Mouse, Clone: FSHb/1062, enQuireBio™
Mouse Monoclonal Antibody
Brand: EnquireBio 2488-MSM1-P1
This item is not returnable.
View return policy
Specifications
| FSH-beta | |
| Cytoplasmic, secreted | |
| FSHb/1062 | |
| Unconjugated | |
| NP_001018090 | |
| Mouse | |
| 22kDa | |
| Protein A/G Purified | |
| RUO | |
| 2488 | |
| IgG1 κ |
| Flow Cytometry, Immunofluorescence, Immunohistochemistry | |
| Monoclonal | |
| 200μg/mL | |
| 10mM PBS with 0.05% BSA and 0.05% azide | |
| FSHB | |
| Recombinant hFSH beta sub-unit | |
| Anterior Pituitary | |
| 100 μg | |
| Primary | |
| Short term: Store at 2-4°C. Long term, aliquot and store at -20°C. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction