missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ FSH beta Monoclonal Antibody (961416)
Rat Monoclonal Antibody
Brand: Invitrogen™ MA561417
This item is not returnable.
View return policy
Description
Reconstitute at 0.5 mg/mL in sterile PBS.
Human follicle stimulating hormone is a glycoprotein produced from urine of post-menopausal women and having a total molecular mass of 35,500 Dalton. HuFSH is a heterodimeric hormone consisting of 165 amino acids a chain (non-active) and 115 amino acids.
Specifications
| FSH beta | |
| Monoclonal | |
| 0.5 mg/mL | |
| PBS with trehalose and no preservative | |
| P01225 | |
| FSHB | |
| E.coli-derived recombinant human FSH B, Asn19-Glu129. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a |
| Western Blot | |
| 961416 | |
| Unconjugated | |
| FSHB | |
| follicle stimulating hormone beta subunit; follicle stimulating hormone, beta polypeptide; follicle-stimulating hormone beta subunit; Follitropin beta chain; follitropin subunit beta; follitropin, beta chain; FSHB; FSH-B; FSH-beta; HH24 | |
| Rat | |
| Protein A/G | |
| RUO | |
| 2488 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Lyophilized |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction