missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FSH beta Antibody (SPM107), mFluor Violet 450 SE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-47735MFV450
This item is not returnable.
View return policy
Description
FSH beta Monoclonal antibody specifically detects FSH beta in Human samples. It is validated for Flow Cytometry
Specifications
| FSH beta | |
| Monoclonal | |
| mFluor Violet 450 SE | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry | |
| SPM107 | |
| Flow Cytometry | |
| follicle stimulating hormone, beta polypeptide, Follicle-stimulating hormone beta subunit, Follitropin beta chain, follitropin subunit beta, follitropin, beta chain, FSH-B, FSH-beta | |
| Full length Purified Human FSH beta (Uniprot: P01225) | |
| 0.1 mL | |
| Cancer | |
| 2488 | |
| Store at 4C in the dark. | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction