missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FSH beta Antibody (961417) [CoraFluor™ 1], Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals FAB43102CL1
This item is not returnable.
View return policy
Description
FSH beta Monoclonal antibody specifically detects FSH beta in Human samples. It is validated for Western Blot
Specifications
| FSH beta | |
| Monoclonal | |
| CoraFluor 1 | |
| follicle stimulating hormone, beta polypeptide, Follicle-stimulating hormone beta subunit, Follitropin beta chain, follitropin subunit beta, follitropin, beta chain, FSH-B, FSH-beta | |
| E.coli-derived recombinant human FSH β, Asn19-Glu129, Accession # P01225 | |
| 0.1 mL | |
| Cancer | |
| 2488 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG2b |
| Western Blot | |
| 961417 | |
| PBS | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction