missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
FSD1 Monoclonal antibody specifically detects FSD1 in Human samples. It is validated for ELISA, Immunoprecipitation, Sandwich ELISA
Specifications
Specifications
| Antigen | FSD1 |
| Applications | ELISA, Immunoprecipitation, Sandwich ELISA |
| Classification | Monoclonal |
| Clone | 1G3 |
| Conjugate | Unconjugated |
| Formulation | In 1x PBS, pH 7.4 |
| Gene Accession No. | BC016442 |
| Gene Alias | fibronectin type 3 and SPRY domain containing 1, fibronectin type III and SPRY domain containing 1, fibronectin type III and SPRY domain-containing protein 1, GLFNDfibronectin type 3 and SPRY (spla, ryanodine) domain containing (withcoiled-coil motif) 1, MGC3213, Microtubule-associated protein GLFND, midline 1-related protein 1, MIR1MID1-related protein 1 |
| Host Species | Mouse |
| Immunogen | FSD1 (-, 1 a.a. ∽ 309 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. MPDEDSKIDHYVLEYRRTNFEGPPRLKEDQPWMVIEGIRQTEYTLTGLKFDMKYMNFRVKACNKAVAGEFSEPVTLETPAFMFRLDASTSHQNLRVDDLSVEWDAMGGKVQDIKAREKDGKGRTASPINSPARGTPSPKRMPSGRGGRDRFTAESYTVLGDTLIDGGEHYWEVRYEPDSKAFGVGVAYRSLGRFEQLGKTAASWCLHVNNWLQVSFTAKHANKVKVLDAPVPDCLGVHCDFHQGLLSFYNARTKQ |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?