Learn More
Invitrogen™ FSCN2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5109738
Description
Immunogen sequence: MPTNGLHQVL KIQFGLVNDT DRYLTAESFG FKVNASAPSL KRKQTWVLEP DPGQGTAVLL RSSHLGRYLS AEEDGRVACE AEQPGRDCRF LVLPQPDGRW VLRSEPHGRF FGGTEDQLSC FATAVSPAEL WTVHLAIHPQ AHLLSVSRRR YVHLCPREDE MAADGDKPWG VDALLTLIFR SRRYCLKSCD SRYLRSDGRL VWEPEPRACY TLEFKAGKLA FKDCDGHYLA PVGPAGTLKA GRNTRPGKDE LFDLEESHPQ VVLVAANHRY VSVRQGVNVS ANQDDELDHE TFLMQIDQET KKCTFYSSTG.
FSCN2 acts as an actin bundling protein. May play a pivotal role in photoreceptor cell-specific events, such as disk morphogenesis.
Specifications
| FSCN2 | |
| Polyclonal | |
| Unconjugated | |
| FSCN2 | |
| A930022G03; Ahl8; C630046B20Rik; fascin 2; fascin 2, retinal; fascin actin-bundling protein 2, retinal; fascin homolog 2, actin-bundling protein, retinal; fascin homolog 2, actin-bundling protein, retinal (Strongylocentrotus purpuratus); fascin-2; Fscn2; Retinal fascin; RFSN; RP30 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 238021, 303741 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 1.27 mg/mL | |
| PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
| Q32M02 | |
| FSCN2 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-310 of human FSCN2 (NP_036550.1). | |
| 100 μL | |
| Primary | |
| Mouse, Rat | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.