missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ FSBP Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5103287
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total FSBP.
Fibrinogen silencer-binding protein, Function: Transcriptional repressor that down-regulates the expression of the fibrinogen gamma chain. Represses transcription of GSK3B gene promoter via its interaction with APBA1. Subunit structure: Interacts with APBA1 (via PDZ 1 and 2 domains). Subcellular location: Nucleus. Tissue specificity: Expressed in multiple tissues including brain. Miscellaneous: Intragenic, in the second intron of RAB54B gene.
Specifications
| FSBP | |
| Polyclonal | |
| Unconjugated | |
| FSBP | |
| fibrinogen silencer binding protein; fibrinogen silencer-binding protein; fibrinogen silencer-binding protein-like; FSBP; LOC102550523 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 100503583, 100861412 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| O95073, Q8BKE5 | |
| FSBP | |
| A synthesized peptide derived from human FSBP(Accession O95073), corresponding to amino acid residues E157-P207. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction