missing translation for 'onlineSavingsMsg'
Learn More

FSBA Antibody (FSBA 2F5/5) - BSA Free, Novus Biologicals™

Product Code. 18408368 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.025 mg
Unit Size:
25µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18408368 0.025 mg 25µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18408368 Supplier Novus Biologicals Supplier No. NB10026560.025mg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

FSBA Monoclonal specifically detects FSBA in Non-species specific samples. It is validated for ELISA, Immunohistochemistry, Immunoprecipitation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FSBA
Applications Western Blot
Classification Monoclonal
Clone FSBA 2F5/5
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot, ELISA 1:100 - 1:2000, Immunohistochemistry 1:10 - 1:500, Immunocytochemistry/Immunofluorescence, Immunoprecipitation 1:10 - 1:500
Gene Alias 5'-(4-(fluorosulfonyl)benzoyl)adenosine, 5'-(4-fluorosulfonylbenzoyl)adenosine, 5'-FSBA, p-fluorosulfonylbenzoyl-5'-adenosine
Host Species Mouse
Immunogen Fluorylsulponylbenzoyladenosine (FSBA)
Purification Method Protein A purified
Quantity 0.025 mg
Regulatory Status RUO
Research Discipline Epitope Tags
Primary or Secondary Primary
Test Specificity FSBA (2F5/5). Chemical.
Target Species Non-species specific
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.