missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ FRYL Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA556643
This item is not returnable.
View return policy
Description
Immunogen sequence: EEVLQIRDET PTLEASLDNA NSRLPEDTTS VLKEEHVTTF EDEGSYIIQE QQESLVCQGI LDLEETEMPE PLAPESYPES VC Highest antigen sequence identity to the following orthologs: Mouse - 80%, Rat - 82%.
FRYL gene ontology annotations related to this gene include cell morphogenesis.
Specifications
| FRYL | |
| Polyclonal | |
| Unconjugated | |
| FRYL | |
| 2010313D22Rik; 2310004H21Rik; 2510002A14Rik; 9030227G01Rik; AF4p12; ALL1-fused gene from chromosome 4p12 protein; DKFZp686E205; FRY like; FRY like transcription coactivator; FRYL; FRY-like; furry homolog-like; furry homolog-like (Drosophila); furry-like; KIAA0826; mKIAA0826; MOR2; mor2 cell polarity protein homolog; protein furry homolog-like; RGD1311309 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 285527 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| O94915 | |
| FRYL | |
| Recombinant protein corresponding to Human FRYL. Recombinant protein control fragment (Product #RP-95479). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction