missing translation for 'onlineSavingsMsg'
Learn More

Fruquintinib O-Desquinazolyl,-O-glucuronide, TRC

Product Code. 30398561
Change view
Click to view available options
Quantity:
5 mg
25 mg
50 mg
Unit Size:
25mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30398561 50 mg 50mg
30407392 25 mg 25mg
30408097 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30398561 Supplier Toronto Research Chemicals Supplier No. TRCF96564050MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Fruquintinib O-Desquinazolyl,-O-glucuronide
Analyte or Component Names Fruquintinib O-Desquinazolyl,-O-glucuronide
Shipping Condition Room Temperature
Molecular Formula C17 H19 N O9
InChI Formula InChI=1S/C17H19NO9/c1-6-10(15(22)18-2)8-4-3-7(5-9(8)25-6)26-17-13(21)11(19)12(20)14(27-17)16(23)24/h3-5,11-14,17,19-21H,1-2H3,(H,18,22)(H,23,24)/t11-,12-,13+,14-,17+/m0/s1
Solution Type Neat
SMILES O[C@@H]1[C@@H](C(O)=O)O[C@@H](OC2=CC=C3C(OC(C)=C3C(NC)=O)=C2)[C@H](O)[C@H]1O
IUPAC Name (2S,3S,4S,5R,6S)-3,4,5-trihydroxy-6-((2-methyl-3-(methylcarbamoyl)benzofuran-6-yl)oxy)tetrahydro-2H-pyran-2-carboxylic acid
Industry Type Pharmaceutical
Molecular Weight (g/mol) 381.34
Quantity 50 mg
Formula Weight 381.11
Visa mer Visa mindre
Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.