missing translation for 'onlineSavingsMsg'
Learn More

Fructosyl Glycine Alpha/Beta Mixture (Mixture of Diastereomers), TRC

Product Code. 17988494
Change view
Click to view available options
Quantity:
10 mg
100 mg
Unit Size:
100mg
10mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17988494 100 mg 100mg
17978764 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17988494 Supplier Toronto Research Chemicals Supplier No. TRCF795750100MG
 Restricted Item
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 60644-20-4
Molecular Formula C8 H15 N O7
Molecular Weight (g/mol) 237.207
Synonym N-(1-Deoxy-β-D-fructopyranos-1-yl)glycine, 1-[(Carboxymethyl)amino]-1-deoxy-β-D-fructopyranose
IUPAC Name 2-[[(3S,4R,5R)-2,3,4,5-tetrahydroxytetrahydropyran-2-yl]methylamino]acetic acid
SMILES O[C@@H]1COC(O)(CNCC(=O)O)[C@@H](O)[C@@H]1O

Specifications

InChI Formula InChI=1S/C8H15NO7/c10-4-2-16-8(15,7(14)6(4)13)3-9-1-5(11)12/h4,6-7,9-10,13-15H,1-3H2,(H,11,12)/t4-,6-,7+,8?/m1/s1
Quantity 100 mg
Formula Weight 237.0849
Percent Purity >95%
Shipping Condition Room Temperature
Chemical Name or Material Fructosyl Glycine Alpha/Beta Mixture (Mixture of Diastereomers)
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.