missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Fructosyl amino acid oxidase Polyclonal antibody specifically detects Fructosyl amino acid oxidase in Bacteria samples. It is validated for Western Blot, ELISA
Specifications
Specifications
| Antigen | Fructosyl amino acid oxidase |
| Applications | Western Blot, ELISA |
| Classification | Polyclonal |
| Conjugate | DyLight 488 |
| Dilution | Western Blot, ELISA |
| Formulation | 50mM Sodium Borate |
| Gene Alias | EC=1.5.3.-, FAOX-C |
| Host Species | Sheep |
| Immunogen | Fructosyl amino acid oxidase [from Corynebacterium sp. Expressed in E.coli] (Uniprot: Q8RIU8) |
| Purification Method | Multi-step |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?